Skip to Content
MilliporeSigma
All Photos(5)

Documents

HPA021453

Sigma-Aldrich

Anti-NPEPPS antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MP100, Anti-PSA, Anti-aminopeptidase puromycin sensitive

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

TLEEARRRFKDHVEGKQILSADLRSPVYLTVLKHGDGTTLDIMLKLHKQADMQEEKNRIERVLGATLLPDLIQKVLTFALSE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The gene NPEPPS (puromycin-sensitive aminopeptidase) is mapped to human chromosome 17q21. The protein mainly localizes in the cytoplasm and nucleus. However, during mitosis it is also present in a membrane-bound form and with the microtubule-spindle apparatus. It is widely expressed in human tissues but strongly expressed in brain, particularly cerebellum.

Immunogen

aminopeptidase puromycin sensitive recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NPEPPS (puromycin-sensitive aminopeptidase) is mainly responsible for amino-terminal removal of antigenic peptides for MHC (major histocompatibility complex) class I antigen presentation and generation of amino acids by breakdown of proteasomal products. It is also suggested to regulate tau-induced pathology in brain by degradation of tau. MiR (microRNA)-614 suppresses cellular proliferation and invasion in lung cancer by silencing NPEPPS.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75648

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunkyung Kim et al.
Journal of immunology (Baltimore, Md. : 1950), 183(11), 7379-7387 (2009-11-18)
Antigenic peptides presented by MHC class I molecules are generated mainly by the proteasome in the cytosol. Several cytosolic aminopeptidases further trim proteasomal products to form mature epitopes or individual amino acids. However, the distinct function of cytosolic aminopeptidases in
N Bhutani et al.
The EMBO journal, 26(5), 1385-1396 (2007-02-24)
Long stretches of glutamine (Q) residues are found in many cellular proteins. Expansion of these polyglutamine (polyQ) sequences is the underlying cause of several neurodegenerative diseases (e.g. Huntington's disease). Eukaryotic proteasomes have been found to digest polyQ sequences in proteins
Fang Lv et al.
Zhongguo fei ai za zhi = Chinese journal of lung cancer, 17(10), 715-721 (2014-10-25)
MicroRNAs (miRNAs) is a group of non-coding small RNA molecules, which play important roles in the development of tumor. The mechanisms of various kinds of miRNAs in lung cancer still need to be further elucidated. This study investigated the function
Soma Sengupta et al.
Biochemistry, 45(50), 15111-15119 (2006-12-13)
Tau, a microtubule associated protein, aggregates into intracellular paired helical filaments (PHFs) by an unknown mechanism in Alzheimer's disease (AD) and other tauopathies. A contributing factor may be a failure to metabolize free cytosolic tau within the neuron. The buildup
W O Bauer et al.
Cytogenetics and cell genetics, 92(3-4), 221-224 (2001-07-04)
Puromycin-sensitive aminopeptidase is a predominantly cytoplasmatic zinc-dependent exopeptidase. Its physiological function is not known to date. Here we report data on tissue distribution, a polymorphism within the coding region and the complete 3' UTR. The gene (NPEPPS alias PSA) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service