Key Documents
GEID2212101
GE Ettan DALT gel cassette
gel size 25 cm × 20 cm
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6067-27
Cytiva ID2212-1,0-1
gel size
25 cm × 20 cm
General description
Rquires no assembly, the two glass plates are attached along one edge by a flexible silicone rubber hinge and the spacers, 1.0- or 1.5-mm thick, are permanently attached to one of the glass plates. No clamps or tape are required in the DALT multi-gel casting unit.
Legal Information
Ettan is a trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)