Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G7640

Sigma-Aldrich

Glycolithocholic acid ethyl ester

Synonym(s):

3α-Hydroxy-5β-cholan-24-oic acid N-(carboxymethyl)amide ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H47NO4
CAS Number:
Molecular Weight:
461.68
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

anionic

SMILES string

[H][C@]12CC[C@]3([H])[C@]([H])(CC[C@]4(C)[C@]([H])(CC[C@@]34[H])[C@H](C)CCC(=O)NCC(=O)OCC)[C@@]1(C)CC[C@@H](O)C2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service