Skip to Content
MilliporeSigma
All Photos(1)

Documents

F7000

Sigma-Aldrich

5-Aminofluorescein

~95% (HPLC)

Synonym(s):

Fluoresceinamine isomer I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13NO5
CAS Number:
Molecular Weight:
347.32
Beilstein/REAXYS Number:
48395
EC Number:
MDL number:
UNSPSC Code:
12171500

assay

~95% (HPLC)

form

powder

color

red-brown

storage temp.

2-8°C

SMILES string

Nc1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35

Looking for similar products? Visit Product Comparison Guide

application

The two isomers are produced by the usual synthesis. Most publications imply they are interchangeable when used for the preparation of the isothiocyanate for fluorescent antibody work. In fact, most commercial preparations are not even labeled as to whether or not they are mixtures.

Other Notes

Note: Do not confuse with fluorescamine.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service