Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

E2877

cis-11,14,17-Eicosatrienoic acid

≥98%, liquid

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C20H34O2
CAS Number:
Molecular Weight:
306.48
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥98%

form

liquid

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/CCCCCCCCCC(O)=O

Packaging

Sealed ampule.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service