agency
meets BP testing specifications, meets EP testing specifications, meets JP testing specifications, meets USP testing specifications
quality
meets EP, BP, JP, USP testing specifications
Gene Information
human ... PYGM(5837)
InChI key
WQZGKKKJIJFFOK-DVKNGEFBSA-N
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
SMILES string
OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
mp
150-152 °C (lit.)
application(s)
pharmaceutical (small molecule)
Looking for similar products? Visit Product Comparison Guide
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service