Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

D6024

Diphenidol hydrochloride

Synonym(s):

1,1-Diphenyl-4-piperidino-1-butanol, Difenidol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C21H27NO · HCl
CAS Number:
Molecular Weight:
345.91
EC Number:
221-850-0
UNSPSC Code:
12352100
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

Cl.OC(CCCN1CCCCC1)(c2ccccc2)c3ccccc3

InChI

1S/C21H27NO.ClH/c23-21(19-11-4-1-5-12-19,20-13-6-2-7-14-20)15-10-18-22-16-8-3-9-17-22;/h1-2,4-7,11-14,23H,3,8-10,15-18H2;1H

InChI key

AVZIYZHXZAYGJS-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service