Skip to Content
MilliporeSigma

CS0200

Prostaglandin EIA Kit

Prostaglandin E2 (PGE2), sufficient for 96 assays

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Prostaglandin EIA Kit, Prostaglandin E2 (PGE2), sufficient for 96 assays

usage

sufficient for 96 assays

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Biological actions include vasodilation, both anti-and pro-inflammatory action, modulation of sleep/wake cycle, stimulation of bone resorption and thermoregulation.
Prostaglandins and thromboxane are products of the cyclooxygenase pathway and are synthesized in response to immediate need and are not stored in significant amounts for later release. Prostanoids mediate various biological functions in humans, including platelet aggregation, gastric mucosa protection, renal homeostasis, vascular homeostasis, uterine function, embryo implantation and labor and sleep/wake cycle.
The sensitivity of this EIA is 15.9 pg/mL and it specifically detects human PGE2 in cell culture supernatants, saliva, urine, plasma and serum without interference from other prostanoids.

Features and Benefits

The assays specifically measure each target lipid with negligible cross-reactivity with 20-25 related eicosanoid compounds tested (from <0.1 to 2%). A standard curve is run in each assay, precoated plates eliminate the coating step, all incubations proceed at room temperature, any commercially available microwell plate reader may be used and reagents are color coded for easy use. The results are available in 4 hours. Performance characteristics show low intra- and inter-assay variability, linear standard curve and recovery of samples ranging from 99-111%.

General description

Competitive enzyme immunoassays (EIA) are employed for the quantitative determination of prostaglandin E1 (PGE1) and prostaglandin E2 (PGE2) in biological fluids. In the assays, the specific protein in the sample competes with a fixed amount of alkaline phosphatase-labeled conjugate for binding sites on the antibody. The intensity of the bound yellow color read at 405 nm is inversely proportional to the concentration of prostanoid in the standards or samples. The measured optical density is used to calculate the concentration.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service