Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS7246

Corning® non-treated 1536 well microplates

black, high base, w/o lid, non-sterile

Synonym(s):

1536 multiwell plates, 1536 well microplates, 1536 well microtiter plates, 1536 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

black
polystyrene plate
square wells

sterility

non-sterile

feature

lid: no

packaging

case of 50 ea

manufacturer/tradename

Corning 7246

size

1536 wells

well volume

12.5 μL

well working volume

1-8 μL

color

black

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service