Documents
CLS48012
Corning® LSE Digital Vortexer Spring Loaded Platfrom for Flasks, Bottles, or Tube Racks
Synonym(s):
Dnase and Rnase free flask, Erlenmeyer flask, conical flasks, flasks
About This Item
UNSPSC Code:
41121804
NACRES:
NB.61
packaging
case of 1 ea
pkg of 1 pkg
manufacturer/tradename
Corning 48012
48012
Related Categories
General description
This is one of three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform has spring loaded retaining bars that adjust to fit vessels of different shapes and sizes.
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.