Skip to Content
MilliporeSigma

C2502

Z-Leu-OH

viscous liquid

Synonym(s):

N-(Benzyloxycarbonyl)-L-leucine, N-(Benzyloxycarbonyl)leucine, N-(Carbobenzoxy)leucine, N-(Phenylmethoxy)carbonyl-L-leucine, N-Carbobenzoxy-L-leucine, N-Carbobenzyloxy-L-leucine, N-Carboxy-L-leucine N-benzyl ester, N-Cbz-L-leucine, Benzyloxycarbonyl-L-leucine, Benzyloxycarbonylleucine, Carbobenzoxyleucine, L-(Carbobenzyloxy)leucine, L-N-(Benzyloxycarbonyl)leucine, NSC 60039, Z-L-Leucine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2CHCH2CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
265.30
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
1253861
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

viscous liquid

refractive index

n20/D 1.512 (lit.)

density

1 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C14H19NO4/c1-10(2)8-12(13(16)17)15-14(18)19-9-11-6-4-3-5-7-11/h3-7,10,12H,8-9H2,1-2H3,(H,15,18)(H,16,17)/t12-/m0/s1

InChI key

USPFMEKVPDBMCG-LBPRGKRZSA-N

Application

Reactant for:
  • Peptide-based preparation of potential antitumor agents
  • Preparation of cyclopropyl peptidomimetics
  • Peptide synthesis




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library