Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
BRAND 26311
Volume:
0.1-200 mL
Feature:
CE compliant
feature
CE compliant
manufacturer/tradename
BRAND 26311
volume
0.1-200 mL
color
magenta
General description
All with one hand: select the delivery mode (gravity-delivery/blow-out) and adjust the motor speed range with your thumb. Perfect fit for glass and plastic pipettes from 0.1 to 200 mL.
Supplied with: A nickel-metal hybride battery, 2 battery compartment covers (place the instrument inverted on the bench), wall support, battery charger (except Z637688 which has no charger) and 2 spare membrane filters 0.2 μm.
- Adjustable motor speed limiter - for slower speed settings allows even more sensitive control of filling and dispensing rates for lower volume pippettes
- Ergonomic handgrip and a weight of only 190 g
- A flashing red LED indicated when the storage battery need charging
- Approx eight hours of continuous pipetting (with a 10 mL pipette) without recharging
- Direct outlet of liquid vapors protects against corrosion to ensure long instrument life
- 0.2 mm hydrophobic membrane filter + built-in check valve protect against liquid penetrating inside
Supplied with: A nickel-metal hybride battery, 2 battery compartment covers (place the instrument inverted on the bench), wall support, battery charger (except Z637688 which has no charger) and 2 spare membrane filters 0.2 μm.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® accu-jet® pro pipette controller
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.