Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

B0501

7-Benzyloxyindole

Synonym(s):

7-(Phenylmethoxy)indole, NSC 92526

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
UNSPSC Code:
12352100
PubChem Substance ID:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C15H13NO/c1-2-5-12(6-3-1)11-17-14-8-4-7-13-9-10-16-15(13)14/h1-10,16H,11H2

SMILES string

C(Oc1cccc2cc[nH]c12)c3ccccc3

InChI key

DIGZMTAFOACVBW-UHFFFAOYSA-N

form

solid

mp

70-74 °C

storage temp.

−20°C

Application

Reactant for preparation of pyrazolo-quinoline derivatives as adenosine receptor antagonists
Reactant for asymmetric total synthesis of diazonamide A
Reactant for preparation of HCV inhibitor
Reactant for preparation of substituted 3-cyanoindoles and 3-(4-pyridinyl)indoles as inhibitors of inosine monophosphate dehydrogenase, with antiproliferative activity on peripheral blood mononuclear cells (PMBC)
Reactant for preparation of [(indolyl)methyl]hydantoin and -thiohydantoin derivatives as treatment of necroptosis
Reactant for preparation of regioisomeric analogs of ED-110, indolocarbazole poisons of human topoisomerase I

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service