Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV40248

Sigma-Aldrich

Anti-SF3B1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Splicing factor 3b, subunit 1, 155 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
guinea pig, dog, rat, human, bovine, rabbit, horse, mouse
citations:
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

87 kDa

species reactivity

guinea pig, dog, rat, human, bovine, rabbit, horse, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SF3B1(23451)

General description

Splicing factor 3b, subunit 1, 155 kDa (SF3B1, SAP155, PRP10) is a subunit of the splicing factor 3b protein complex, which along with splicing factor 3a and 12S RNA forms the U2 small nuclear ribonucleoproteins complex (U2 snRNP). SF3B1 is also a component of the minor U12-type spliceosome. Mutations of SF3B1have been linked to myelodysplastic syndromes (MDS) and chronic lymphocytic leukemia (CLL).

Specificity

Anti-SF3B1 (AB1) polyclonal antibody reacts with zebrafish, chicken, canine, human, mouse, and rat splicing factor 3b, subunit 1, 155 kDa proteins.

Immunogen

Synthetic peptide directed towards the middle region of human SF3B1

Application

Anti-SF3B1 (AB1) polyclonal antibody is used to tag splicing factor 3b, subunit 1, 155 kDa for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of splicing factor 3b, subunit 1, 155 kDa in U2-snRNP and U12-type spliceosome formation and function.

Biochem/physiol Actions

SF3B1 is the subunit 1 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex (U2 snRNP). The splicing factor 3b/3a complex binds pre-mRNA upstream of the intron′s branch site in a sequence independent manner and may anchor the U2 snRNP to the pre-mRNA. Splicing factor 3b is also a component of the minor U12-type spliceosome.

Sequence

Synthetic peptide located within the following region: LLNDIPQSTEQYDPFAEHRPPKIADREDEYKKHRRTMIISPERLDPFADG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service