Skip to Content
MilliporeSigma
All Photos(1)

Documents

A214600

Sigma-Aldrich

DMT-2′O-TC-rA(ib) Phosphoramidite

Synonym(s):

DMT-2′O-TC-rA(ib) Amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H61N8O10PS2
CAS Number:
Molecular Weight:
1017.16
UNSPSC Code:
41116105

product line

Proligo Reagents

Quality Level

assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

technique(s)

oligo synthesis: suitable

impurities

≤-0.5% 31P-NMR P(III) impurites 145-155ppm (31P-NMR)
≤-0.5% Impurity 3′-TC Isomer (reversed phase HPLC)

nucleoside profile

base: adenosine
base protecting group: isobutyryl
2' protecting group: TC
5' protecting group: DMT

storage temp.

-10 to -25°C

SMILES string

S=C(N1CCS(CC1)(=O)=O)O[C@@H]2[C@H](OP(N(C(C)C)C(C)C)OCCC#N)[C@H](O[C@H]2N3C4=C(N=C3)C(NC(C(C)C)=O)=NC=N4)COC(C5=CC=C(OC)C=C5)(C6=CC=C(OC)C=C6)C7=CC=CC=C7

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service