Skip to Content
MilliporeSigma
All Photos(1)

Documents

40559

Sigma-Aldrich

3-Piperonylcyclobutanone

≥98.0% (HPLC)

Synonym(s):

3-(1,3-Benzodioxol-5-ylmethyl)cyclobutanone, 3-(3,4-Methylenedioxybenzyl)cyclobutanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12O3
CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥98.0% (HPLC)

form

liquid

SMILES string

O=C1CC(C1)Cc2ccc3OCOc3c2

InChI

1S/C12H12O3/c13-10-4-9(5-10)3-8-1-2-11-12(6-8)15-7-14-11/h1-2,6,9H,3-5,7H2

InChI key

AWAOUJUBBHDZRM-UHFFFAOYSA-N

Application

3-Piperonylcyclobutanone may be used as a reagent in the chiral synthesis of lignan lactones such as (-)-hinokinin, (-)-deoxypodorhizone, (-)-isohibalactone and (-)-savinin.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral synthesis of lignan lactones, (?)-hinokinin, (?)-deoxypodorhizone, (?)-isohibalactone and (?)-savinin by means of enantioselective deprotonation strategy.
Toshio Honda, Nobuaki Kimura,
Journal of the Chemical Society. Perkin Transactions 1, 1, 1043-1046 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service