Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

29469

N-Cyclohexyl-N′-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate

≥99.0% (CHN)

Synonym(s):

N-Cyclohexyl-N′-(β-[N-methylmorpholino]ethyl)carbodiimide p-toluenesulfonate, CMC, CME-CDI

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C14H26N3O · C7H7O3S
CAS Number:
Molecular Weight:
423.57
EC Number:
219-650-3
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
3872192
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

GBCAVSYHPPARHX-UHFFFAOYSA-M

InChI

1S/C14H26N3O.C7H8O3S/c1-17(9-11-18-12-10-17)8-7-15-13-16-14-5-3-2-4-6-14;1-6-2-4-7(5-3-6)11(8,9)10/h14H,2-12H2,1H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.C[N+]2(CCOCC2)CCN=C=NC3CCCCC3

assay

≥99.0% (CHN)

ign. residue

≤0.1% (as SO4)

mp

115-120 °C

Application

Water-soluble condensing reagent for peptide synthesis, reagent used for coupling ligands to carriers in affinity chromatography.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Hui Wang et al.
Journal of hepatology, 63(2), 370-377 (2015-03-24)
The asialoglycoprotein receptor on hepatocyte membranes recognizes the galactose residues of glycoproteins. We investigated the specificity, accuracy and threshold value of asialoglycoprotein receptor imaging for estimating liver reserve via scintigraphy using (111)In-hexavalent lactoside in mouse models. (111)In-hexavalent lactoside scintigraphy for
Renata A Castro et al.
SpringerPlus, 3, 382-382 (2014-08-12)
The mangrove ecosystem is a coastal tropical biome located in the transition zone between land and sea that is characterized by periodic flooding, which confers unique and specific environmental conditions on this biome. In these ecosystems, the vegetation is dominated
Yongjun Wei et al.
PloS one, 10(6), e0129921-e0129921 (2015-06-13)
Diverse cellulolytic bacteria are essential for maintaining high lignocellulose degradation ability in biogas digesters. However, little was known about functional genes and gene clusters of dominant cellulolytic bacteria in biogas digesters. This is the foundation to understand lignocellulose degradation mechanisms
Yubiao Liu et al.
Chemical communications (Cambridge, England), (4)(4), 398-399 (2004-02-07)
Poly(N,N-dialkylcarbodiimide) was found to be an effective polymeric ligand system for preparing and stabilizing palladium nanoparticles (1-5 nm). The composite material prepared in situ was found to be a robust catalyst for the Suzuki coupling reaction under microwave or regular
Katherine A Kellersberger et al.
Analytical chemistry, 76(9), 2438-2445 (2004-05-01)
A top-down approach based on sustained off-resonance irradiation collision-induced dissociation (SORI-CID) has been implemented on an electrospray ionization (ESI) Fourier transform mass spectrometer (FTMS) to characterize nucleic acid substrates modified by structural probes. Solvent accessibility reagents, such as dimethyl sulfate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service