Z803634
Hellma® cylindrical absorption cuvettes
High Performance Quartz Glass, spectral range 200-2,500 nm, pathlength 100 mm, chamber volume 28,000 μL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
Suprasil® quartz
description
with 2 polished windows
manufacturer/tradename
Hellma 120-QS
parameter
200-2,500 nm spectral range
technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
D
102.5 mm
chamber volume
28,000 μL
pathlength
100 mm
detection method
photometric
Looking for similar products? Visit Product Comparison Guide
General description
with 2 stoppers
Packaging
Sold as packages containing a single cell (1EA = 1 cell)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service