Key Documents
Z800813
Hellma® reference plate for qualifying microplate readers
to check photometric accuracy and wavelength accuracy
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
manufacturer/tradename
Hellma (666-R113)
application(s)
food and beverages
petroleum
pharmaceutical
plastics
General description
Material: Neutral Density Glass Filter NG 5 (0.5), NG 4 (1.0), NG 3 (1.5), (2.0), Holmium Oxide glass filter
Wavelength: 405, 450, 490, 650, 279, 361, 453, 536, 638
With reference plates you can check the photometric and wavelength accuracy of microplate readers. They have the same dimensions as a microplate with 96 wells and a 6.6 mm diameter per window (height 14.5 x width 125 x length 85.5 mm).
Application
Hellma® reference plate for qualifying microplate readers as a calibration standard for checking the photometric and wavelength accuracy of microplate readers.
Features and Benefits
  • Accredited to DAkkS Deutsche Akkreditierungsstelle (National accreditation body for the Federal Republic of Germany)
  • Traceable to NIST (National Institute of Standards and Technology)
  • Assured Quality of the measurement results
  • Proven accuracy and reliability
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)