Skip to Content
MilliporeSigma
All Photos(2)

Documents

PHR8633

Supelco

Ethyl (2S)2-[(3S,5aS,8aS, 9aS)-3-methyl-1,4-dioxodecahydro-1H-cyclopenta[e]pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoate

certified reference material, pharmaceutical secondary standard

Synonym(s):

Ramipril diketopiperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N2O4
CAS Number:
Molecular Weight:
398.50
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

ramipril

packaging

pkg of 50MG

application(s)

pharmaceutical

InChI

1S/C23H30N2O4/c1-3-29-23(28)19(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-18-11-7-10-17(18)14-20(25)22(24)27/h4-6,8-9,15,17-20H,3,7,10-14H2,1-2H3/t15-,17-,18-,19-,20-/m0/s1

InChI key

KOVMAAYRBJCASY-JBDAPHQKSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service