Skip to Content
MilliporeSigma
All Photos(1)

Documents

CCW903

Millipore

Plate Count Agar Plates

new

Medium for aerobic plate counts by the surface inoculation method (standard Plate Count Agar) according to ISO 4833, 8552 & 17410 Standards and IFU No. 6, suitable for microbiology

Synonym(s):

Aerobic Plate count Agar, Casein-peptone Dextrose Yeast Agar, PCA, Standard Methods Agar, Tryptone Glucose Yeast Extract Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606

agency

IFU 6
ISO 17410
ISO 4833
ISO 8552

Quality Level

form

solid

shelf life

3.5 mo. at 2‑14 °C (Expiry date on the label)

feature

ready-to-use

composition

agar, 15.0 g/L
casein, 5.00 g/L
dextrose, 1.00 g/L
yeast extract, 2.50 g/L

packaging

box of 20 plates

storage condition

(Store dry and tightly closed)

parameter

21 mL filling volume

plate diam.

9 mm

color

yellowish

pH

7.0±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

nonselective for

General description

The Plate Count Agar formulation is modified to avoid the addition of milk. The new composition allows the growth of most microorganisms without any further additions. The medium′s formulation is equivalent to that described by the ‘Standard Methods for the Examination of Dairy products’, the USP′s ‘Tryptone Glucose Yeast Agar’, the ‘Deutsche Landswirtchaft’ and the APHA, ISO, and AOAC′s Plate Count Agar. This is the medium of choice for the plate count of any type of sample.

Application

The Plate Count Agar is suitable for aerobic plate counts by the surface inoculation method (standard Plate Count Agar).

Components

  • 1 box with 2 packs of 10 plates/pack
  • Single cellophane

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service