BCR178
Calcium ammonium nitrate fertilizer (composition)
BCR®, certified reference material
Synonym(s):
Calcium ammonium nitrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
agency
BCR®
manufacturer/tradename
JRC
application(s)
agriculture
environmental
format
matrix material
Analysis Note
For more information please see:
BCR178
BCR178
Legal Information
BCR is a registered trademark of European Commission
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service