Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94292

Supelco

Abberior® STAR 488, biotin

≥90.0% (HPCE)

Sign Into View Organizational & Contract Pricing


About This Item

assay

≥90.0% (HPCE)

form

liquid

manufacturer/tradename

Abberior GmbH

fluorescence

λex 504 nm; λem 524 nm±10 nm in DMSO

detection method

fluorometric

storage temp.

2-8°C

General description

Abberior® STAR 488 dye was developed for STED and confocal microscopy in the green spectral region. It can be very effectively excited with the prominent 488 nm laser line. For STED microscopy, this fluorescent dye can be efficiently used with STED laser wavelengths between 590 - 610 nm. This category of dyes can substitute dyes like Oregon Green 488, Atto 488 or ALEXA FLUOR 488. Organic fluorescent dye conjugate with biotin is suitable for fluorescence labeling of avidin- or streptavidin-modified molecules.

Features and Benefits

  • Combine highest brightness and photostability.
  • Excellent solubility in aqueous buffers as PBS.
  • Allows background-free labeling in cells or tissue.

Suitability

Designed and tested for fluorescent super-resolution microscopy

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
Atto is a trademark of Atto-Tec GmbH
abberior is a registered trademark of Abberior GmbH

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service