89891
cis-1,2-Cyclohexanecarboxylic anhydride
analytical standard
Synonym(s):
cis-Cyclohexane-1,2-dicarboxylic acid anhydride, cis-Hexahydro-1,3-isobenzofurandione, cis-Hexahydrophthalic anhydride
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
storage temp.
2-8°C
SMILES string
[H][C@]12[C@](C(OC2=O)=O)([H])CCCC1
InChI
1S/C8H10O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h5-6H,1-4H2
InChI key
MUTGBJKUEZFXGO-UHFFFAOYSA-N
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service