Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76655

Sigma-Aldrich

Pentaerythritol triacrylate

technical, ~50% (GC)

Synonym(s):

PETA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCO2CH2)3CCH2OH
CAS Number:
Molecular Weight:
298.29
Beilstein/REAXYS Number:
6598752
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical

concentration

~50% (GC)

refractive index

n20/D 1.483 (lit.)
n20/D 1.483

density

1.18 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

InChI

1S/C14H18O7/c1-4-11(16)19-8-14(7-15,9-20-12(17)5-2)10-21-13(18)6-3/h4-6,15H,1-3,7-10H2

InChI key

HVVWZTWDBSEWIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suk-Yong Jang et al.
Scientific reports, 6, 38050-38050 (2016-12-20)
Currently, Si as an active material for LIBs has been attracting much attention due to its high theoretical specific capacity (3572 mAh g-1). However, a disadvantage when using a Si negative electrode for LIBs is the abrupt drop of its capabilities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service