Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32109

Supelco

Prothipendyl-(dimethyl-d6) oxalate

VETRANAL®, analytical standard

Synonym(s):

10-{3-[Di(methyl-d3)amino]propyl}-10H-pyrido[3,2-b][1,4]benzothiazine oxalate salt, N,N-Di(methyl-d3)-3-pyrido[3,2-b][1,4]benzothiazin-10-ylpropan-1-amine oxalate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D6H13N3S · C2H2O4
Molecular Weight:
381.48
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)C(O)=O.CN(C)CCCN1c2ccccc2Sc3cccnc13

General description

Prothipendyl-(dimethyl-d6) oxalate is a high purity veterinary drug standard used for the determination of the active ingredients of the drug in processed food of animal origin.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Guo et al.
Molecules (Basel, Switzerland), 26(17) (2021-09-11)
A novel class of styryl sulfones were designed and synthesized as CAPE derivatives by our work team, which showed a multi-target neuroprotective effect, including antioxidative and anti-neuroinflammatory properties. However, the underlying mechanisms remain unclear. In the present study, the anti-Parkinson's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service