Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OF-0005

Oxyrase

Oxyrase Enzyme System in OxyFluor

1x5mL, Sterile 30u/ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$70.39

$70.39


Available to ship onMarch 25, 2025Details



Select a Size

Change View
1 EA
$70.39

About This Item

PDF Icon
Shipped & Sold By Oxyrase
A Trusted Marketplace Seller
PDF Icon
Have a question?

$70.39


Available to ship onMarch 25, 2025Details


biological source

Escherichia coli

storage temp.

-20 °C

storage condition

avoid repeated freeze/thaw cycles

manufacturer/tradename

Oxyrase

General description

OxyFluor™ was formulated to meet the needs of the microscopist using fluorescent dyes.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library