OA-0100
Oxyrase
Oxyrase for Agar
1x100mL, Sterile 30u/ml
Select a Size
$105.26
Available to ship onMarch 26, 2025Details
Select a Size
About This Item
$105.26
Available to ship onMarch 26, 2025Details
Recommended Products
biological source
Escherichia coli
storage temp.
-20 °C
storage condition
avoid repeated freeze/thaw cycles
manufacturer/tradename
Oxyrase
General description
Oxyrase® for Agar (OA) is a sterile enzyme formulation that utilizes the Oxyrase® Enzyme System to produce anaerobic conditions in your custom agar.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.