Select a Size
About This Item
Skip To
material
polyethersulfone membrane, polystyrene
sterility
sterile
feature
holdup volume 2.4 mL, hydrophilic
manufacturer/tradename
Stericup®
parameter
150 mL process volume, 150 mL sample volume, 45 °C max. temp.
technique(s)
sterile filtration: suitable
H
120 mm
diam.
12.5 cm
matrix
Express PLUS™
pore size
0.22 μm nominal pore size, 0.22 μm pore size
fitting
inlet funnel, outlet flask (Receiver)
filter capacity
150 mL
filter diam.
73 mm
filtration area
40 cm2
shipped in
ambient
storage temp.
room temp
General description
Features & Benefits
Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications
Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component
Biochem/physiol Actions
Physical form
Preparation Note
Gamma irradiation
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
"Precise cell assays start with Stericup® media filtration. Poor cell health and erratic treatment responses are often caused by culturing inconsistencies. Reduce your culture variability by using Stericup® filters with Millipore Express® PLUS membranes and realize rapid sterile filtration with minimal clogging, protein binding or loss of material."
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service