Skip to Content
MilliporeSigma

MXRP2USBC

Milliflex® Rapid 2.0 Detection Tower – USB cable


Sign In to View Organizational & Contract Pricing.

Select a Size

This product is sold exclusively by Millipore via offline channels. Contact Customer Service

About This Item

NACRES:
NA.74
UNSPSC Code:
41122409

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

input

sample type pharmaceutical(s)

application(s)

bioburden testing
pharmaceutical
sterility testing

compatibility

for use with Milliflex® Rapid System 2.0

shipped in

ambient

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MXRP2ADAPMXRDP2DT00MXRDP2KT01
compatibility

for use with Milliflex® Rapid System 2.0

compatibility

for use with Milliflex® Rapid System 2.0

compatibility

for use with Milliflex® Rapid System 2.0

compatibility

for use with Milliflex Oasis®

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

-

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

pkg of 1 kit

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

-

technique(s)

Rapid Sterility Testing: suitable

technique(s)

Rapid Sterility Testing: suitable

technique(s)

Rapid Sterility Testing: suitable

technique(s)

Rapid Sterility Testing: suitable

input

sample type pharmaceutical(s)

input

-

input

sample type pharmaceutical(s)

input

-

Application

Rapid detection; response and resolution of microbial contamination in Pharmaceutical Products Analysis

General description

The Milliflex® Rapid System 2.0 is an automated solution for the rapid detection, imaging, and quantification of viable microbial contaminants in filterable samples throughout the manufacturing process. Its results help to improve process control, product yield and the timely release of final products. In case of contamination, corrective action can be taken earlier, avoiding loss of time, money, and production capacity

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

User guide on Milliflex Rapid 2 Detection Tower

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service