Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE1115M

Sigma-Aldrich

Anti-Pax1 Antibody, clone 5A2

clone 5A2, from rat

Synonym(s):

Paired box protein Pax-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A2, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Pax1(18503)

General description

Paired box protein Pax-1 (UniProt: P09084) is encoded by the Pax1 (also known as Pax-1) gene (Gene ID: 18503) in murine species. Pax-1 is a transcriptional activator that plays a role in the formation of segmented structures of the embryo and has important role in the normal development of the vertebral column. Mutations in Pax1 gene are associated with vertebral malformations. Undulated homozygous mice exhibits vertebral malformations along the entire rostro-caudal axis. Pax genes are vital for the specification of progenitor cells and for maintenance of progenitor cell fate, including that of cancer cells. Pax1 gene is silenced by methylation in ovarian and cervical cancers and may serve as a tumor suppressor gene. The methylation levels of Pax1 gene is significantly higher in tumor tissues compared to non-tumor paracancerous tissues. Hence, methylation status of Pax1 may serve as useful biomarkers for the detection of cervical cancer and is also a good prognostic indicator for oral squamous cell carcinoma. (Ref.: Huang J et al (2017). Int. J. Environ. Res. Public Health. 14(2). pii: E216).

Specificity

Clone 5A2 detects Paired box protein Pax-1 in murine tissues.

Immunogen

GST-tagged full-length murine recombinant paired box protein Pax-1.

Application

Anti-Pax1, clone 5A2, Cat. No. MABE1115, is a highly specific rat monoclonal antibody that targets Paired box protein Pax-1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Western Blotting Analysis: A representative lot detected Pax1 in Western Blotting applications (Feederle, R., et. al. (2016). Monoclon Antib Immunodiagn Immunother. [Epub ahead of print]).

Immunohistochemistry Analysis: A representative lot detected Pax1 in Immunohistochemistry applications (Feederle, R., et. al. (2016). Monoclon Antib Immunodiagn Immunother. [Epub ahead of print]; Kist, R., et. al. (2014). PLoS Genet. 10(10):e1004709).

Immunohistochemistry Analysis: A 1:200 dilution from a representative lot detected Pax1 in Embryonic mouse head (E14.5) (Courtesy of Dr. Heiko Peters, Ph.D., Institute of Genetic Medicine, Newcastle University, United Kingdom).

Western Blotting Analysis: A 1:10 dilution from a representative lot detected Pax1 in d11.5 embryonic vertebral column tissue lysates (Courtesy of Dr. Heiko Peters, Ph.D., Institute of Genetic Medicine, Newcastle University, United Kingdom).

Quality

Evaluated by Western Blotting in mouse esophagus tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Pax1 in 10 µg of mouse esophagus tissue lysate.

Target description

~46 kDa observed; 46.26 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Yu et al.
eLife, 12 (2024-01-26)
Adolescent idiopathic scoliosis (AIS) is a common and progressive spinal deformity in children that exhibits striking sexual dimorphism, with girls at more than fivefold greater risk of severe disease compared to boys. Despite its medical impact, the molecular mechanisms that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service