Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CE-125ML-RU

Millipore

Anti-Fya from the clone P3TIM, Research Use Only

Anti-Fyafrom the clone P3TIM, Human IgG, For non-IVD/Research Use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

human

Quality Level

antibody form

unpurified

clone

P3TIM, monoclonal

form

liquid

packaging

pkg of 125 mL

technique(s)

agglutination assay: suitable

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

General description

Clarified cell culture supernatant from cell lines expressing Human IgG monoclonal blod typing antibodies. For non-IVD/ research use. Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.

Application

Bulk for non IVD/Research Use
For Non-IVD / Research use only

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1%(w/v) sodium azide.

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service