Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AX1697A

Supelco

Coulomat A

for use with AQUASTAR®

Synonym(s):

Coulometric Anode Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601

form

liquid

Quality Level

compatibility

for use with AQUASTAR®

shipped in

ambient

storage temp.

room temp

General description

Coulomat A is an AQUASTAR® coulometric reagent mixture for use in Karl Fischer titration to determine the moisture content in samples with a high degree of accuracy.

Application

AX1697A-1 and AX1697A-30 are packaged in 38/430 poly coated glass bottles.
AQUASTAR® Coulomat A reagent is used to determine the water content of bitumen and residual samples by Karl Fischer titration.(1)

Analysis Note

Color: Colorless to light yellow
Form: Liquid
Performance: To pass test
Water (H2O): 1-50ppm

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 Oral - STOT RE 2 Oral - STOT SE 1 - STOT SE 3

target_organs

Central nervous system, Eyes,Central nervous system, Liver,Kidney, Thyroid

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Reliable determination of water contents of bitumen and vacuum residua via coulometric Karl Fischer titration using tetrahydrofuran
Carbognani L, et al.
Petroleum Science and Technology, 32(5), 602-609 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service