Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

ABC425

Anti-Trop-2 Antibody

from rabbit

Synonym(s):

Tumor-associated calcium signal transducer 2, Cell surface glycoprotein Trop-2, Membrane component chromosome 1 surface marker 1, Pancreatic carcinoma marker protein GA733-1

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TACSTD2(4070)

General description

The protein known as Trop-2, or also known as cell surface glycoprotein Trop-2, Membrane component chromosome 1 surface marker 1 or Pancreatic carcinoma marker protein GA733-1 and encoded by the gene TACSTD2/GA733-1/M1S1/TROP2, is a protein that may function as a growth factor receptor and is expressed primarily in the placenta, lung and epithelial tissues but also found expressed in pancreatic cancer cell lines. Interestingly, this protein′s gene is intronless, a rare event in eukaryotic genes, and was first identified as a carcinoma antigen by the monoclonal GA733. It is a plasma membrane bound protein that transduces an intracellular calcium signal and appears to act as a receptor for as yet unidentified growth factors. Interestingly too, mutations in this gene are associated with a rare cause of blindness with amyloid protein bodies (insoluble fibrous protein aggregates but not always of amyloid protein) beneath the cornea. Trop-2 has also recently been shown to be important in regulating fibroblast motility and lung cell proliferation and does so by regulating the ERK kinase pathway as well as several critical components of the cell migration apparatus.
~ 36 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: Intracellular domain
Linear peptide corresponding to the intracellular domain of human Trop-2.

Application

Anti-Trop-2 Antibody detects level of Trop-2 & has been published & validated for use in Trop-2.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Trop-2 in 10 µg of HT1376, SiHa, A549, TERA-2, T47D, OVCAR 3, Colo-587, PANC-1, JEG-3, HCC1143, DU145, NT2/D1 , SK-OV-3, LNCap cell lysate.
Immunohistochemistry Analysis: A representative lot detected Trop-2 in in human prostate cancer tissues (Stoyanova, T., et al. (2012). Genes Dev. 26:2271-2285).

Physical form

Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in Tera-2 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Trop-2 in 10 µg of Tera-2 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlin A Kowalsky et al.
The Journal of biological chemistry, 290(44), 26457-26470 (2015-08-25)
Knowledge of the fine location of neutralizing and non-neutralizing epitopes on human pathogens affords a better understanding of the structural basis of antibody efficacy, which will expedite rational design of vaccines, prophylactics, and therapeutics. However, full utilization of the wealth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service