8.1887A
Lithium aluminium hydride
(powder) for synthesis
Synonym(s):
Lithium aluminium hydride, LAH, Lithium alanate, Aluminium lithium hydride
About This Item
Recommended Products
Quality Level
form
powder
mp
125 °C (decomposition)
density
0.917 g/cm3 at 20 °C
bulk density
400 kg/m3
InChI
1S/Al.Li.4H/q-1;+1;;;;
InChI key
OCZDCIYGECBNKL-UHFFFAOYSA-N
Application
Analysis Note
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1A - Water-react. 1
Storage Class
4.3 - Hazardous materials, which set free flammable gases upon contact with water
wgk_germany
WGK 1
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service