Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72103

Millipore

ProteoExtract® Glycopeptide Enrichment Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Protein glycosylation influences crucial biological and pharmaceutically relevant events. Glycoanalysis studies of antibodies, receptors and other glycosylated proteins require highly sensitive, selective, efficient and reproducible methods for sample preparation and mass spectrometry (MS). Effective analysis requires glycopeptide enrichment to compensate for low abundance glycopeptides in complex mixtures and, subsequently, to enable glycan and attached peptide analysis by MS.The ProteoExtract Glycopeptide Enrichment Kit is designed to capture high-mannose, hybrid, and complex type glycosylated peptides from complex protein mixtures. The kit uses a proprietary ZIC Glycocapture Resin. In combination with the included optimized buffers, the ZIC Glycocapture Resin is highly selective for glycosylated peptides in the presence of abundant non-glycosylated peptides. The protocol is optimized to allow efficient removal of non-glycosylated peptides with an almost complete glycopeptide recovery (as quantitatively assessed by N-Glycan mapping). It allows both site-specific determination of the glycan structure and parallel analysis of the attached peptides. Glycopeptide enrichment is non-specific and nonselective, without preference for particular glycan structures and without loss of distinct glycans. The enriched glycopeptide fraction is directly compatible with downstream analysis by MALDI and ESI-MS. One ProteoExtract Glycopeptide Enrichment Kit provides sufficient reagents for 50 enrichment reactions. 10 µl sample with total protein concentration 2-4 mg/ml.

Warning

Toxicity: Flammable (J)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEOEXTRACT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service