Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.00502

Sigma-Aldrich

Proteasome Inhibitor XXI, Z-LLLL-SF

Synonym(s):

Proteasome Inhibitor XXI, Z-LLLL-SF, Cbz-Leu-Leu-Leu-Leu-ψ-[CH2SO2]-F, Cbz-Leu₄-SF, Cbz-LLLL-SF, Z-Leu₄-SF, Z-L₄-SF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H53FN4O7S
CAS Number:
Molecular Weight:
656.85
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable tetrapeptidylsulfonylfluoride-based irreversible proteasome inhibitor that selectively inhibits the hydrolysis of Suc-LLVY-AMC (Cat. No. 539142) by the β5 chymotrypsin-like activity (IC50 = 7 nM using yeast 20S proteasome; [E]0 = 0.1 nM, [S]0 = 250 µM; 15 min preincubation before substrate addition), while exhibiting much reduced potency against Z-LLE-AMC (Cat. No. 539141) hydrolysis by the β1 caspase-like activity or Z-LLR-AMC hydrolysis by the β2 trypsin-like activity (29% and 75% inhibition, respectively; 15 min preincubation with 200 µM inhibitor). One hour inhibitor treatment at 5 µM in HEK cultures is shown to completely abolish β5 labeling by the proteasome probe MV151, while only partial MV151 blockage is seen with β1 and β2 subunits at an inhibitor concentration of 50 µM.
A cell-permeable tetrapeptidylsulfonylfluoride-based irreversible proteasome inhibitor that selectively inhibits the hydrolysis of Suc-LLVY-AMC (Cat. No. 539142) by the β5 chymotrypsin-like activity (IC50 = 7 nM using yeast 20S proteasome), while exhibiting much reduced potency against Z-LLE-AMC (Cat. No. 539141) hydrolysis by the β1 caspase-like activity or Z-LLR-AMC hydrolysis by the β2 trypsin-like activity (29% and 75% inhibition, respectively, with 200 µM inhibitor). One hour inhibitor treatment at 5 µM in HEK cultures is shown to completely abolish β5 labeling by the proteasome probe MV151, while only partial MV151 blockage is seen with β1 and β2 at an inhibitor concentration of 50 µM.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
β5 chymotrypsin-like activity of 20S proteasome
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Brouwer. A.J., et al. 2013. J. Med. Chem.55, 10995.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service