Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

292500

Sigma-Aldrich

Diethylamine NONOate

A nitric oxide (NO) donor that releases NO in aqueous solutions at controlled rates.

Synonym(s):

Diethylamine NONOate, DEA NONOate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H22N4O2
CAS Number:
Molecular Weight:
206.29
UNSPSC Code:
12352200

Quality Level

assay

≥98% (elemental analysis)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

wet ice

storage temp.

−70°C

General description

A nitric oxide (NO) donor that releases NO in aqueous solutions at controlled rates.
Reagent that releases nitric oxide (NO) in aqueous solutions at controlled rates. The half-life (t½) of NO release = 16 min (PBS, pH 7.4, 22°C).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A nitric oxide (NO) donor that releases NO in aqueous solutions.
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Ramamurthi, A., and Lewis, R.S. 1997. Chem. Res. Toxicol. 10, 408.
Laval, F., and Wink, D.A. 1994. Carcinogenesis 5, 443.
Maragos, C.M., et al. 1993. CancerRes. 53, 564.
Morley, D., and Keefer, L.K. 1993. J. Cardiovasc. Pharmacol.22, S3.
Wink, D.A., et al. 1993. Proc. Natl. Acad. Sci. USA90, 9813.
Maragos, C.M., et al. 1991. J. Med. Chem.34, 3242.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service