Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

230752

Millipore

Chromomycin A3

Antitumor antibiotic that inhibits RNA synthesis. A cell-permeable fluorescent dye that can be used with Bisbenzimide H 33258 to distinguish chromosomes by their total DNA content and DNA base composition.

Synonym(s):

Chromomycin A3, Toyomycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H82O26
CAS Number:
Molecular Weight:
1183.25
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

ethanol: 10 mg/mL

fluorescence

λex ~445 nm
λem ~575 nm

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C57H82O26/c1-21-34(79-40-19-37(53(26(6)75-40)77-28(8)59)82-38-16-33(61)52(70-11)25(5)74-38)15-31-13-30-14-32(54(71-12)51(68)46(63)22(2)58)55(50(67)44(30)49(66)43(31)45(21)62)83-41-18-35(47(64)24(4)73-41)80-39-17-36(48(65)23(3)72-39)81-42-20-57(10,69)56(27(7)76-42)78-29(9)60/h13,15,22-27,32-33,35-42,46-48,52-56,58,61-66,69H,14,16-20H2,1-12H3/t22-,23-,24-,25-,26-,27+,32+,33-,35-,36-,37-,38-,39+,40+,41+,42+,46+,47-,48-,52+,53+,54+,55+,56+,57+/m1/s1

InChI key

ZYVSOIYQKUDENJ-WKSBCEQHSA-N

General description

Antibiotic that inhibits RNA synthesis by way of DNA-dependent RNA polymerase. Fluorescent dye that can be used in conjunction with Bisbenzimide H33258 (Cat. No. 382061) to distinguish chromosomes by their total DNA concentration and their DNA base composition. Binds to G:C pairs of helical DNA (Mg2+ required) but does not intercalate the A:T pairs, which preferentially bind Bisbenzimide H33258.
Antitumor antibiotic that inhibits RNA synthesis. A cell-permeable fluorescent dye that can be used with Bisbenzimide H 33258 (Cat. No. 382061) to distinguish chromosomes by their total DNA content and DNA base composition. Reversibly interacts with DNA in the presence of Mg2+. Binds to G:C pairs of helical DNA but does not intercalate.

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Reconstitution

Following recosntitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Aich, P., et al. 1992. Biochemistry31, 2988.
Saccone, S., et al. 1992. Proc. Natl. Acad. Sci. USA89, 4913.
Van Dilla, M.A., et al. 1983. Science220, 620.
Mizuno, K. 1963. J. Antibiot.16, 22.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service