Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.44251

Millipore

Universal Rapid Battery Charger, 120 V (US)

for use with RCS® High Flow/{TM="RCS"] Plus Ex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow/{TM="RCS"] Plus Ex

storage temp.

room temp

General description

940371 (RCS High Flow/RCS Plus Ex)
For charging the 9.6 V and 7.2 V Ni-Cd batteries used with the RCS High Flow and instruments of the RCS Plus series (RCS Plus, RCS Plus Ex), respectively.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service