Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.10646

Supelco

Reagent bottles, 100 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24122003

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

no temp limit

General description

We offer a big variety of packaging materials like empty bottles and containers for your own preparations and mixtures of our reagents.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service