Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09879

Supelco

Buffer concentrate for buffer solution acc. to WEISE

for 1000 ml of solution (phosphate), pH 7.20 Titrisol®

Synonym(s):

Weise′s Buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161706
NACRES:
NA.25

grade

analytical standard

Quality Level

agency

according to DIN 19266

product line

Titrisol®

pH

7.20 (20 °C in H2O)

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The Titrisol® reference buffer concentrate for buffer solution acc. to WEISE [for 1000 mL of solution (phosphate), pH=7.20] is analyzed in the ISO/IEC 17025 accredited calibration laboratory with a combined glass electrode and 5-point calibration according to DIN 19268 against a certified primary standard reference material following DIN 19266.

The Titrisol® concentrated buffer material combines ease of use and maximum precision for the routine calibration and monitoring of pH instruments. To prepare the ready-to-use pH buffer solution, simply dilute the Titrisol® buffer concentrate.

Application


  • Anthocyanin content analysis: A study highlighted the influence of molar absorbance on the anthocyanin content of foods, utilizing Petunidin 3-glucoside chloride as a standard in analytical methods to ensure accurate quantification in various food matrices. This application is crucial for researchers focusing on food chemistry and polyphenol biochemistry (Chandra Singh et al., 2022).

  • Oxygen radical scavenging capacity (ORAC) in cereals: Research on the anthocyanin composition and ORAC of cereals employed Petunidin 3-glucoside chloride to examine its antioxidant properties in purple, black, and common barley, supporting studies in plant pigment analysis and the health impacts of flavonoid compounds (Bellido and Beta, 2009).

Features and Benefits

  • Traceable to primary standard reference materials from NIST and PTB
  • Analytical standard Grade
  • Uncertainty data and traceability are documented in a comprehensive, batch-specific Certificate of Analysis.

Analysis Note

pH-value (20 °C): 7.20 +/- 0.05
Measured after dilution to 1l at 20 °C.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service