Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01562

Supelco

Absorption tube for CO₂

(sodium hydroxide on support)

Synonym(s):

Carbon dioxide absorption

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104911

Quality Level

pH

13.8 (20 °C, 50 g/L in H2O)

bulk density

800 kg/m3

storage temp.

2-30°C

Analysis Note

Absorption capacity for CO₂: ≥ 50.0 %

pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk_germany

WGK 1

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service