Skip to Content
MilliporeSigma
All Photos(1)

Documents

07-209-I

Sigma-Aldrich

Anti-phospho-SHC Antibody (Tyr239, Tyr240)

from rabbit, purified by affinity chromatography

Synonym(s):

p46Shc (pTyr194/pTyr195), p52Shc (pTyr239/pTyr240), p66Shc (pTyr349/pTyr350), SHC-transforming protein 1, tyrosine-phosphorylated, SH2 domain protein C1, tyrosine-phosphorylated, SHC-transforming protein 3, tyrosine-phosphorylated, SHC-transforming prote

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), bovine (based on 100% sequence homology), rat (based on 100% sequence homology), chicken (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pTyr239/pTyr240)

Gene Information

human ... SHC1(6464)

General description

SHC-transforming protein 1 (UniProt P29353; also known as SH2 domain protein C1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1) is encoded by the SHC1 (also known as SHC, SHCA) gene (Gene ID 6464) in human. Originally identified as an SH2-containing proto-oncogene involved in growth factor signaling, Shc proteins function as adaptor molecules recruited by numerous receptors (e.g. integrins, GPCRs, and receptors for growth factors, antigens, cytokines, and hormones) for signling. In addition to the ubiquitously expressed Shc (a.k.a. ShcA by SHC1/SHCA gene), there exist also ShcB (by SHC2/SHCB gene) and ShcC (by SHC3/SHCC gene), which are predominantly expressed in neuronal cells. Alternative splicings result in six ShcA isoforms, including p66Shc, p52Shc, and p46Shc. All ShcA isoforms possess two distinct domains, PTB (a.a. 156-339) and SH2 (a.a. 488-579), that bind phosphotyrosine-containing sequences, as well as a central region (CH1; a.a. 340-487) that contains critical tyrosine phosphorylation sites. The phosphotyrosine-binding (PTB) domain is also referred to as phosphotyrosine-interaction domain or PID.

Specificity

Numbering of the target pTyr residues varies depending on the species and spliced isoforms. Target pTyr residues this rabbit polyclonal detects correspond to pTyr349/pTyr350 of human spliced isoform 6 and p66Shc, pTyr239/pTyr240 of human spliced isoform 7 and p52Shc, pTyr194/pTyr195 of isoform p46Shc, and pTyr135/pTyr136 of isoform 5 reported by UniProt (P29353).

Immunogen

KLH-conjugated linear peptide corresponding to a target site sequence surrounding phosphorylated Tyr239 and Tyr240 of human spliced isoform p52Shc.

Application

Detect SHC tyr239 and tyr240 phosphorylation using this rabbit polyclonal Anti-phospho-SHC (Tyr239, Tyr240) antiboody, Cat. No. 07-209-I, validated for use in Western Blotting.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.
Western Blotting Analysis: 2 µg/mL of this antibody detected SHC tyrosine phosphorylation induction in 10 µg of lysate from EGF-stimulated HEK293 cells.

Target description

~52 kDa observed. 62.82/51.61/46.67/40.42/62.89/51.68 kDa (human isoform p66Shc/p52Shc/p46Shc/5/6/7), 62.61/51.41/46.47 kDa (mouse isoform p66Shc/p52Shc/p47Shc), and 51.50 kDa (rat) calculated.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service