Skip to Content
MilliporeSigma
All Photos(2)

Documents

07-1477

Sigma-Aldrich

Anti-Rsk1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

90 kDa ribosomal protein S6 kinase 1, MAP kinase-activated protein kinase 1, Ribosomal S6 kinase 1, Ribosomal protein S6 kinase alpha-1, S6K-alpha 1, p90-RSK 1, Ribosomal protein S6 kinase alpha 1, Ribosomal protein S6 kinase, 90kD, 1, Ribosmal protein S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RPS6KA1(6195)

General description

Rsk1, Rsk2, and Rsk3 (ribosomal protein S6 kinase, 90kDa, polypeptide 1,2,3; S6K-alpha 1,2,3) are Ser/Thr kinases which are activated by MAP Kinases, and are alternatively referred to as MAPKAP kinases 1a, 1b, and 1c. Following activation, Rsk isoforms translocate to the nucleus, where Rsk2 phosphorylates Ser10 of Histone H3, a critical step in the stimulation of immediate-early gene expression. Rsk1 is able to phosphorylate Ser133 of CREB, activating it as a transcription factor. Rsk1 activation can be monitored by phosphorylation state specific antibodies directed against sites involved in its activation.

Specificity

Recognizes p90 RSK1.

Immunogen

Synthetic peptide corresponding to the c-terminal sequence of human p90 RSK1 (C-LAQRRVRKLPSTTL).

application

Anti-Rsk1 Antibody is an antibody against Rsk1 for use in WB, IP, ELISA.
Immunoprecipitation: 4 µg of a previous lot of this antibody was used per 1500 µg of crude protein sample in Immunoprecipitation.

ELISA: Recommended.
Optimal working dilutions must be determined by end user.

Western Blot: A previous lot of this antibody was tested on multiple lysates. See datasheet.

Quality

Routinely evaluated by Western Blot on NIH/3T3 lysates.

Western Blot Analysis:
1:500-1:1,000 dilution of this lot detected RSK1 on 10 μg of NIH/3T3 lysates.

Target description

83 kDa

Linkage

Replaces: 04-417

Physical form

Purified rabbit polyclonal IgG in buffer containing TBS with 0.01% Sodium Azide in 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C in undiluted aliquots from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerolcontaining solutions to become frozen during storage.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pricing of drugs and donations: options for sustainable equity pricing.
C Perez-Casas,E Herranz,N Ford
Tropical Medicine & International Health null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service