Key Documents
HS3900
Hausser Scientific
Hausser Scientific™ Petroff-Hausser Bacteria Counter
Counting chamber with two cover glasses
Synonym(s):
hemacytometer, counting chamber
Select a Size
1 EA
$705.00
$705.00
Available to ship onApril 02, 2025Details
Select a Size
Change View
1 EA
$705.00
About This Item
PDF Icon
Shipped & Sold By HAUSSER SCIENTIFIC CO.
A Trusted Marketplace Seller
PDF Icon
Have a question?
$705.00
Available to ship onApril 02, 2025Details
manufacturer/tradename
Hausser Scientific
General description
The Petroff-Hausser Counter is one piece construction ensuring durability and accuracy, featuring an Improved Neubauer ruling on a single plateau. The Petroff-Hausser Counting Chamber is popular for bacteria and sperm counting and offered in a series of cell-depths. This chamber is 20 microns deep.
Choose from one of the most recent versions:
Certificates of Analysis (COA)