Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1509013

Foxx Life Sciences

Foxx Life Sciences Borosil® Reagent Bottles Amber

Amber Reagent Bottles Plain Narrow Mouth, Graduated I/C Glass Stopper 60 mL, 40/CS

Synonym(s):

Bottles

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?

capacity

60 mL

feature

with cap

material

Bottle: borosilicate glass 3.3
Cap: plastic

manufacturer/tradename

Foxx Life Sciences

General description

Borosil® Amber Reagent Bottles, graduated with interchangeable amber flat head stoppers are chemically resistant and mechanically strong. These narrow mouth storage vessels have a sturdy ground neck and come standard with graduations and an enameled area for easy marking. The amber color is highly durable and protects light-sensitive media from light radiation. All components are autoclavable and sterilizable. High resistance to thermal shock up to 140°C. Complies with IS 1388 (Part II), EN ISO 4796 - 2. *60mL & 125mL not covered in IS and EN ISO standards.


SPECIFICATIONS

  • Material: Glass
  • Bottle: 60 mL
  • Overall Dimension: 40mm X 100mm
  • Neck Type: Grounded Neck I/C Stopper
  • Borosilicate Glass 3.3
  • Graduation Range: Type I
  • Quantity: Case of 40

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library