Skip to Content
MilliporeSigma
All Photos(1)

Documents

ENA444723232

Enamine

3-nitro-4-sulfamoylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
246.2
MDL number:

mp

260-261

Iupac Name

3-nitro-4-sulfamoylbenzoic acid

Inchi Code

1S/C7H6N2O6S/c8-16(14,15)6-2-1-4(7(10)11)3-5(6)9(12)13/h1-3H,(H,10,11)(H2,8,14,15)

InChI key

CVNYDIJDUSXBSC-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-144249

Melting Point

260-261

MSDS

https://www.enaminestore.com/catalog/EN300-144249


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service