Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA341294752

Enamine

2-(1-ethyl-1H-imidazol-2-yl)acetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
SEK 3,980.88
100 MG
SEK 5,685.18
250 MG
SEK 7,893.36
500 MG
SEK 12,131.88
1 G
SEK 15,421.92
2.5 G
SEK 29,738.04
5 G
SEK 43,757.76
10 G
SEK 64,653.96

Vendor SKU: EN300-5353687-50MG

SEK 3,980.88


Availability
Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
SEK 3,980.88
100 MG
SEK 5,685.18
250 MG
SEK 7,893.36
500 MG
SEK 12,131.88
1 G
SEK 15,421.92
2.5 G
SEK 29,738.04
5 G
SEK 43,757.76
10 G
SEK 64,653.96

About This Item

CAS Number:
Molecular Weight:
135.17
MDL number:

Vendor SKU: EN300-5353687-50MG

SEK 3,980.88


Availability
Ships in 14 days - from Aldrich Partner.

Iupac Name

(1-ethyl-1H-imidazol-2-yl)acetonitrile

Inchi Code

1S/C7H9N3/c1-2-10-6-5-9-7(10)3-4-8/h5-6H,2-3H2,1H3

InChI key

LDTYEUBXAVXINI-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-90318

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-90318


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service