Key Documents
CH196032
Eisco Labs
Eisco Labs Glass Erlenmeyer Flask Set
Glass Erlenmeyer Flask Set - 100ml, 250ml & 500ml - Narrow Neck, White Graduations
Synonym(s):
flask set, Glass Flasks, Erlenmeyer Flasks
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • INDUSTRIAL STRENGTH || Made of high quality, heavy duty, Borosilicate 3.3 glass. Thick, uniform walls provide excellent strength against breakage. Designed for use in professional laboratories, but is ideal for classroom, garage and home use
  • 3 PIECE SET || Set includes three Erlenmeyer flasks. 100ml, 250ml and 500ml capacities. Flasks feature narrow necks, beaded rims and white, printed graduations
  • SUPERIOR HEAT and CHEMICAL DURABILITY || Microwavable and autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks
  • SAFETY PACK || Set is packaged in a custom fit, thick foam casing, for safe storage and worry free transport
  • MANUFACTURED BY EISCO LABS || Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind
Three piece Erlenmeyer Flask set made of high quality, heavy duty, borosilicate 3.3 glass. Includes three, narrow neck flasks with 100ml, 250ml and 500ml capacities. Flasks feature white, printed graduations, and beaded rims. Uniform, thick walls provide excellent strength against breakage. Set is packaged in a custom fit, thick foam casing, for safe storage and worry free transport.
Measurements:
[ 100ml ] 4.2" tall, 2.6" wide base. Neck, 21mm inner diameter, 26mm outer diameter.
[ 250ml ] 5.7" tall, 3.3" wide base. Neck, 29mm inner diameter, 34mm outer diameter.
[ 500ml ] 7" tall, 4.25" wide base. Neck, 29mm inner diameter, 34mm outer diameter.
Microwavable and autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco Labs Borosilicate glass has the following heat tolerances: Strain Point -515°C | Annealing Point 565°C | Softening Point 820°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)